CD170 (Siglec F) Monoclonal Antibody (1RNM44N), eFluor™ 660, eBioscience™ - FAQs
CD170 (Siglec F) Monoclonal Antibody (1RNM44N), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD170 (Siglec F) Monoclonal Antibody (1RNM44N), eFluor™ 660, eBioscience™ - FAQs (50-1702-82, 50-1702-80)