CD177 Monoclonal Antibody (MEM-166 (MEM166)), eFluor™ 660, eBioscience™ - FAQs
CD177 Monoclonal Antibody (MEM-166 (MEM166)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD177 Monoclonal Antibody (MEM-166 (MEM166)), eFluor™ 660, eBioscience™ - FAQs (50-1779-42, 50-1779-41)