CD183 (CXCR3) Monoclonal Antibody (CEW33D), eFluor™ 660, eBioscience™ - FAQs
CD183 (CXCR3) Monoclonal Antibody (CEW33D), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD183 (CXCR3) Monoclonal Antibody (CEW33D), eFluor™ 660, eBioscience™ - FAQs (50-1839-41, 50-1839-42)