CD199 (CCR9) Monoclonal Antibody (BBC3M4), eFluor™ 660, eBioscience™ - FAQs
CD199 (CCR9) Monoclonal Antibody (BBC3M4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD199 (CCR9) Monoclonal Antibody (BBC3M4), eFluor™ 660, eBioscience™ - FAQs (50-1999-42, 50-1999-41)