CD209a Monoclonal Antibody (MMD3), eFluor™ 660, eBioscience™ - FAQs
CD209a Monoclonal Antibody (MMD3), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD209a Monoclonal Antibody (MMD3), eFluor™ 660, eBioscience™ - FAQs (50-2094-80, 50-2094-82)