beta Catenin Monoclonal Antibody (15B8), eFluor™ 660, eBioscience™ - FAQs
beta Catenin Monoclonal Antibody (15B8), eFluor™ 660, eBioscience™ - FAQs
View additional product information for beta Catenin Monoclonal Antibody (15B8), eFluor™ 660, eBioscience™ - FAQs (50-2567-42, 50-2567-41)