CD294 (CRTH2) Monoclonal Antibody (BM16), eFluor™ 660, eBioscience™ - FAQs
CD294 (CRTH2) Monoclonal Antibody (BM16), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD294 (CRTH2) Monoclonal Antibody (BM16), eFluor™ 660, eBioscience™ - FAQs (50-2949-42, 50-2949-41)