CD300c Monoclonal Antibody (TX45), eFluor™ 660, eBioscience™ - FAQs
CD300c Monoclonal Antibody (TX45), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD300c Monoclonal Antibody (TX45), eFluor™ 660, eBioscience™ - FAQs (50-3006-42)