CD300f (IREM-1) Monoclonal Antibody (UP-D1), eFluor™ 660, eBioscience™ - FAQs
CD300f (IREM-1) Monoclonal Antibody (UP-D1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD300f (IREM-1) Monoclonal Antibody (UP-D1), eFluor™ 660, eBioscience™ - FAQs (50-3008-41, 50-3008-42)