CXCL9 (MIG) Monoclonal Antibody (MIG-2F5.5), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CXCL9 (MIG) Monoclonal Antibody (MIG-2F5.5), eFluor™ 660, eBioscience™ - FAQs (50-3009-80)