CD301b (MGL2) Monoclonal Antibody (11A10-B7), eFluor™ 660, eBioscience™ - FAQs
CD301b (MGL2) Monoclonal Antibody (11A10-B7), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD301b (MGL2) Monoclonal Antibody (11A10-B7), eFluor™ 660, eBioscience™ - FAQs (50-3011-82, 50-3011-80)