CD307e (FcRL5) Monoclonal Antibody (509F6), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD307e (FcRL5) Monoclonal Antibody (509F6), eFluor™ 660, eBioscience™ - FAQs (50-3078-42, 50-3078-41)