Pokemon (LRF) Monoclonal Antibody (13E9), eFluor™ 660, eBioscience™ - FAQs
Pokemon (LRF) Monoclonal Antibody (13E9), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Pokemon (LRF) Monoclonal Antibody (13E9), eFluor™ 660, eBioscience™ - FAQs (50-3309-80, 50-3309-82)