CD335 (NKp46) Monoclonal Antibody (29A1.4), eFluor™ 660, eBioscience™ - FAQs
CD335 (NKp46) Monoclonal Antibody (29A1.4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD335 (NKp46) Monoclonal Antibody (29A1.4), eFluor™ 660, eBioscience™ - FAQs (50-3351-80, 50-3351-82)