Jagged 2 Monoclonal Antibody (HMJ2-1), eFluor™ 660, eBioscience™ - FAQs
Jagged 2 Monoclonal Antibody (HMJ2-1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Jagged 2 Monoclonal Antibody (HMJ2-1), eFluor™ 660, eBioscience™ - FAQs (50-3392-80)