CD354 (TREM-1) Monoclonal Antibody (Lndnb3r), eFluor™ 660, eBioscience™ - FAQs
CD354 (TREM-1) Monoclonal Antibody (Lndnb3r), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD354 (TREM-1) Monoclonal Antibody (Lndnb3r), eFluor™ 660, eBioscience™ - FAQs (50-3549-42)