Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs
Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs
View additional product information for Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs (50-4015-82, 50-4015-80)