F(ab')2-Goat anti-Rat IgG (H+L) Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs
View additional product information for F(ab')2-Goat anti-Rat IgG (H+L) Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs (50-4017-82)