Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 660, eBioscience™ - FAQs (50-4301-82, 50-4301-80)