Product Image
Invitrogen™

eBioscience™ Streptavidin eFluor™ 660 Conjugate

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
50-4317-8025 μg
50-4317-82100 μg
Catalog number 50-4317-80
Price (USD)
181.00
Each
Quantity:
25 μg
Price (USD)
181.00
Each

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

Reported Applications

  • Flow Cytometric Analysis
  • Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
  • Immunohistochemical Staining of Frozen Tissue Sections
  • Immunocytochemistry

For Research Use Only.

Specifications
Concentration0.2 mg/mL
Excitation/Emission633-647/668 nm
Flow Cytometer Laser LinesRed
Product TypeStreptavidin Conjugate (fluorescent)
Quantity25 μg
Target MoleculeBiotin
ConjugateeFluor 660
FormLiquid
Product LineeBioscience
Unit SizeEach

Frequently asked questions (FAQs)

Is the eFluor 660 fluorochrome compatible with Anti-Cy5/Alexa Fluor 647 beads?

Yes, in-house studies have demonstrated that the eFluor 660 fluorochrome is recognized by Anti-Cy5/Alexa Fluor 647 beads. Side by side studies with Alexa Fluor 647 versus eFluor 660 conjugated antibodies have demonstrated comparable results.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.