eBioscience™ Streptavidin eFluor™ 660 Conjugate, 100 μg - FAQs
eBioscience™ Streptavidin eFluor™ 660 Conjugate, 100 μg - FAQs
View additional product information for eBioscience™ Streptavidin eFluor™ 660 Conjugate - FAQs (50-4317-82, 50-4317-80)