Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 660, eBioscience™ - FAQs
Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 660, eBioscience™ - FAQs (50-4321-80, 50-4321-82)