Rat IgM Isotype Control (eBRM), eFluor™ 660, eBioscience™ - FAQs
Rat IgM Isotype Control (eBRM), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Rat IgM Isotype Control (eBRM), eFluor™ 660, eBioscience™ - FAQs (50-4341-82, 50-4341-80)