Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 660, eBioscience™ - FAQs
Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 660, eBioscience™ - FAQs (50-4724-80)