Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 660, eBioscience™ - FAQs
Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 660, eBioscience™ - FAQs (50-4732-82, 50-4732-80)