Mouse IgM Isotype Control (11E10), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Mouse IgM Isotype Control (11E10), eFluor™ 660, eBioscience™ - FAQs (50-4752-82, 50-4752-80)