FOXP3 Monoclonal Antibody (150D/E4), eFluor™ 660, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (150D/E4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (150D/E4), eFluor™ 660, eBioscience™ - FAQs (50-4774-41, 50-4774-42)