FOXP3 Monoclonal Antibody (PCH101), eFluor™ 660, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (PCH101), eFluor™ 660, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (PCH101), eFluor™ 660, eBioscience™ - FAQs (50-4776-41, 50-4776-42)