FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 660, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 660, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 660, eBioscience™ - FAQs (50-4777-41, 50-4777-42)