FOXA2 Monoclonal Antibody (3C10), eFluor™ 660, eBioscience™ - FAQs
FOXA2 Monoclonal Antibody (3C10), eFluor™ 660, eBioscience™ - FAQs
View additional product information for FOXA2 Monoclonal Antibody (3C10), eFluor™ 660, eBioscience™ - FAQs (50-4778-80, 50-4778-82)