F4/80 Monoclonal Antibody (BM8), eFluor™ 660, eBioscience™ - FAQs
F4/80 Monoclonal Antibody (BM8), eFluor™ 660, eBioscience™ - FAQs
View additional product information for F4/80 Monoclonal Antibody (BM8), eFluor™ 660, eBioscience™ - FAQs (50-4801-80, 50-4801-82)