Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs
Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs
View additional product information for Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs (50-4817-80, 50-4817-82)