EOMES Monoclonal Antibody (Dan11mag), eFluor™ 660, eBioscience™ - FAQs
EOMES Monoclonal Antibody (Dan11mag), eFluor™ 660, eBioscience™ - FAQs
View additional product information for EOMES Monoclonal Antibody (Dan11mag), eFluor™ 660, eBioscience™ - FAQs (50-4875-80, 50-4875-82)