EOMES Monoclonal Antibody (WD1928), eFluor™ 660, eBioscience™ - FAQs
EOMES Monoclonal Antibody (WD1928), eFluor™ 660, eBioscience™ - FAQs
View additional product information for EOMES Monoclonal Antibody (WD1928), eFluor™ 660, eBioscience™ - FAQs (50-4877-41, 50-4877-42)