Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 660, eBioscience™ - FAQs
Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 660, eBioscience™ - FAQs (50-4888-82, 50-4888-80)