phospho-Tyrosine Monoclonal Antibody (pY20), eFluor™ 660, eBioscience™ - FAQs
phospho-Tyrosine Monoclonal Antibody (pY20), eFluor™ 660, eBioscience™ - FAQs
View additional product information for phospho-Tyrosine Monoclonal Antibody (pY20), eFluor™ 660, eBioscience™ - FAQs (50-5001-41, 50-5001-42)