CD200 Monoclonal Antibody (OX90), eFluor™ 660, eBioscience™ - FAQs
CD200 Monoclonal Antibody (OX90), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD200 Monoclonal Antibody (OX90), eFluor™ 660, eBioscience™ - FAQs (50-5200-80, 50-5200-82)