CD8b Monoclonal Antibody (SIDI8BEE), eFluor™ 660, eBioscience™ - FAQs
CD8b Monoclonal Antibody (SIDI8BEE), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD8b Monoclonal Antibody (SIDI8BEE), eFluor™ 660, eBioscience™ - FAQs (50-5273-41, 50-5273-42)