3G11 Sialoganglioside Antigen Monoclonal Antibody (eBio3G11 (SM3G11, 3G11)), eFluor™ 660, eBioscience™ - FAQs
3G11 Sialoganglioside Antigen Monoclonal Antibody (eBio3G11 (SM3G11, 3G11)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for 3G11 Sialoganglioside Antigen Monoclonal Antibody (eBio3G11 (SM3G11, 3G11)), eFluor™ 660, eBioscience™ - FAQs (50-5443-80, 50-5443-82)