Ki-67 Monoclonal Antibody (SolA15), eFluor™ 660, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (SolA15), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (SolA15), eFluor™ 660, eBioscience™ - FAQs (50-5698-80, 50-5698-82)