Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 660, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 660, eBioscience™ - FAQs (50-5699-80, 50-5699-82)