TCR V delta 4 Monoclonal Antibody (GL2), eFluor™ 660, eBioscience™ - FAQs
TCR V delta 4 Monoclonal Antibody (GL2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for TCR V delta 4 Monoclonal Antibody (GL2), eFluor™ 660, eBioscience™ - FAQs (50-5702-80)