GPNMB Monoclonal Antibody (CTSREVL), eFluor™ 660, eBioscience™ - FAQs
GPNMB Monoclonal Antibody (CTSREVL), eFluor™ 660, eBioscience™ - FAQs
View additional product information for GPNMB Monoclonal Antibody (CTSREVL), eFluor™ 660, eBioscience™ - FAQs (50-5708-80, 50-5708-82)