CD169 (Siglec-1) Monoclonal Antibody (SER-4), eFluor™ 660, eBioscience™ - FAQs
CD169 (Siglec-1) Monoclonal Antibody (SER-4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD169 (Siglec-1) Monoclonal Antibody (SER-4), eFluor™ 660, eBioscience™ - FAQs (50-5755-82, 50-5755-80)