Eos Monoclonal Antibody (ESB7C2), eFluor™ 660, eBioscience™ - FAQs
Eos Monoclonal Antibody (ESB7C2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Eos Monoclonal Antibody (ESB7C2), eFluor™ 660, eBioscience™ - FAQs (50-5758-80)