FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 660, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 660, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 660, eBioscience™ - FAQs (50-5773-80, 50-5773-82)