CD85k (Gp49b) Monoclonal Antibody (H1.1), eFluor™ 660, eBioscience™ - FAQs
CD85k (Gp49b) Monoclonal Antibody (H1.1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD85k (Gp49b) Monoclonal Antibody (H1.1), eFluor™ 660, eBioscience™ - FAQs (50-5784-82)