Aiolos Monoclonal Antibody (8B2), eFluor™ 660, eBioscience™ - FAQs
Aiolos Monoclonal Antibody (8B2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Aiolos Monoclonal Antibody (8B2), eFluor™ 660, eBioscience™ - FAQs (50-5789-80)