T-bet Monoclonal Antibody (eBio4B10 (4B10)), eFluor™ 660, eBioscience™ - FAQs
T-bet Monoclonal Antibody (eBio4B10 (4B10)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for T-bet Monoclonal Antibody (eBio4B10 (4B10)), eFluor™ 660, eBioscience™ - FAQs (50-5825-80, 50-5825-82)