OCT3/4 Monoclonal Antibody (EM92), eFluor™ 660, eBioscience™ - FAQs
OCT3/4 Monoclonal Antibody (EM92), eFluor™ 660, eBioscience™ - FAQs
View additional product information for OCT3/4 Monoclonal Antibody (EM92), eFluor™ 660, eBioscience™ - FAQs (50-5841-80, 50-5841-82)